(R)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A3308-1G A3308-25G A3308-5G
TCA3308-1GEA 44 USD
TCA3308-1G TCA3308-25G TCA3308-5G
(R)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
(R)-3-Aminobutyric acid
Formula: C₄H₉NO₂
MW: 103.1198 g/mol
Melting Pt: 212°C
MDL Number: MFCD00211284
CAS Number: 3775-73-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR