Tetrabutylammonium toluene-4-sulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

T3660-25G T3660-5G
TCT3660-25GEA 231 USD
TCT3660-25G TCT3660-5G
Tetrabutylammonium toluene-4-sulfonate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium toluene-4-sulphonate
Formula: C₂₃H₄₃NO₃S
MDL Number: MFCD00043227
CAS Number: 7182-86-7
EINECS: 230-548-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR