2,4-Dimethoxy-3-methylbenzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

D5789-25G D5789-5G
TCD5789-25GEA 303 USD
TCD5789-25G TCD5789-5G
2,4-Dimethoxy-3-methylbenzaldehyde ≥98.0% (by GC)
2,4-Dimethoxy-3-methylbenzaldehyde
Formula: C₁₀H₁₂O₃
Boiling Pt: 109°C
MDL Number: MFCD00011489
CAS Number: 7149-92-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR