(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
A2782-1G
A2782-5G
TCA2782-1GEA
47
USD
TCA2782-1G
TCA2782-5G
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) ) ≥98.0% (by HPLC)
(7-Azabenzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (AOP) )
Formula:
C₁₂H₂₂F₆N₆OP₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09263289 CAS Number: 156311-85-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Lambda max. | 245.0 to 248.0 nm (CH3CN) |
Absorbance | min. 0.280 (0.02g/L, CH3CN, 245.0 to 248.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...