4-Methoxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Methoxy-3-methylbenzeneboronic acid

M2809-1G M2809-5G
TCM2809-1GEA 45 USD
TCM2809-1G TCM2809-5G
4-Methoxy-3-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Methoxy-3-methylphenylboronic acid
Formula: C₈H₁₁BO₃
MDL Number: MFCD02179464
CAS Number: 175883-62-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 98.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR