2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

D4120-25G D4120-5G
TCD4120-25GEA 151.7 USD
TCD4120-25G TCD4120-5G
2-Amino-3,4-dimethylbenzoic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-3,4-dimethylbenzoic acid
Formula: C₉H₁₁NO₂
MDL Number: MFCD00130041
CAS Number: 50419-58-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR