4'-Hydroxyacetophenone oxime ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
H1512-1G
H1512-5G
TCH1512-1GEA
64
USD
TCH1512-1G
TCH1512-5G
4'-Hydroxyacetophenone oxime ≥98.0% (by GC)
4'-Hydroxyacetophenone oxime
Formula:
C₈H₉NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00481697 CAS Number: 34523-34-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...