4'-(Difluoromethoxy)acetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

D4760-1G D4760-5G
TCD4760-1GEA 84.7 USD
TCD4760-1G TCD4760-5G
4'-(Difluoromethoxy)acetophenone ≥98.0% (by GC)
4'-(Difluoromethoxy)acetophenone
Formula: C₉H₈F₂O₂
MDL Number: MFCD00042250
CAS Number: 83882-67-1
EINECS: 281-174-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR