3'-Brom-4'-methyloacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3'-Bromo-4'-methylacetophenone, 1-(3-Bromo-4-methylphenyl)ethanone
B4758-1G
B4758-5G
TCB4758-1GEA
28
USD
TCB4758-1G
TCB4758-5G
3'-Brom-4'-methyloacetophenone ≥98.0% (by GC)
3'-Brom-4'-methyloacetophenone
Formula:
C₉H₉BrO MW: 213.0700 g/mol Boiling Pt: 118°C |
MDL Number:
MFCD02683854 CAS Number: 40180-80-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...