4-Benzyloxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Benzyloxy-3-chlorobenzeneboronic acid, (4-(Benzyloxy)-3-chlorophenyl)boronic acid

B5064-1G B5064-5G
TCB5064-1GEA 36 USD
TCB5064-1G TCB5064-5G
4-Benzyloxy-3-chlorophenylboronic acid (contains varying amounts of Anhydride)
4-Benzyloxy-3-chlorophenylboronic acid
Formula: C₁₃H₁₂BClO₃
MDL Number: MFCD04115642
CAS Number: 845551-44-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) gt. 97.0 and le 108.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR