2'-Hydroxy-4',5'-dimethylacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4812-1G
D4812-5G
TCD4812-1GEA
38
USD
TCD4812-1G
TCD4812-5G
2'-Hydroxy-4',5'-dimethylacetophenone ≥98.0% (by GC)
2'-Hydroxy-4',5'-dimethylacetophenone
Formula:
C₁₀H₁₂O₂ Boiling Pt: 144°C |
MDL Number:
MFCD00002306 CAS Number: 36436-65-4 EINECS: 253-035-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...