5-Pyrimidineboronic acid pinacol ester ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Pyrimidine-5-boronic acid pinacol ester, Pyrimidyl-5-boronic acid pinacol ester

T2521-1G T2521-5G
TCT2521-1GEA 48 USD
TCT2521-1G TCT2521-5G
5-Pyrimidineboronic acid pinacol ester ≥96.0% (by GC, titration analysis)
5-Pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₅BN₂O₂
MDL Number: MFCD05155223
CAS Number: 321724-19-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR