4-Bromo-1,3,5-trimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Bromo-1,3,5-trimethylpyrazole

B4744-1G B4744-5G
TCB4744-1GEA 35 USD
TCB4744-1G TCB4744-5G
4-Bromo-1,3,5-trimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
4-Bromo-1,3,5-trimethyl-1H-pyrazole
Formula: C₆H₉BrN₂
MW: 189.06
Boiling Pt: 97°C
MDL Number: MFCD00052532
CAS Number: 15801-69-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR