Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)

Supplier: TCI America
Danger

Synonyms: Tetradecyltrihexylphosphonium dicyanamide

T3534-25G T3534-5G
TCT3534-25GEA 153 USD
TCT3534-25G TCT3534-5G
Trihexyl(tetradecyl)phosphonium dicyanamide ≥93.0% (by total nitrogen basis)
Trihexyl(tetradecyl)phosphonium dicyanamide
Formula: C₃₄H₆₈N₃P
MW: 549.90 g/mol
MDL Number: MFCD04117699
CAS Number: 701921-71-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (with Total Nitrogen) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR