1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3560-25G T3560-5G
TCT3560-25GEA 290 USD
TCT3560-25G TCT3560-5G
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane ≥97.0% (by GC)
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Formula: C₁₁H₁₃F₁₃O₃Si
Boiling Pt: 71°C
MDL Number: MFCD00153261
CAS Number: 85857-16-5
EINECS: 288-657-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR