5-Chloro-2-hydroxy-4-methylbenzophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3260-25G C3260-5G
TCC3260-25GEA 230 USD
TCC3260-25G TCC3260-5G
5-Chloro-2-hydroxy-4-methylbenzophenone ≥98.0% (by GC)
5-Chloro-2-hydroxy-4-methylbenzophenone
Formula: C₁₄H₁₁ClO₂
MDL Number: MFCD00002325
CAS Number: 68751-90-6
EINECS: 272-140-2

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR