3-Iodo-1H-pyrazole ≥97.0% (by GC)

Supplier: TCI America
Warning

I1016-1G I1016-5G
TCI1016-1GEA 101 USD
TCI1016-1G TCI1016-5G
3-Iodo-1H-pyrazole ≥97.0% (by GC)
3-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR