3',5'-Dichloro-2'-hydroxyacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

D4742-25G D4742-5G
TCD4742-25GEA 221 USD
TCD4742-25G TCD4742-5G
3',5'-Dichloro-2'-hydroxyacetophenone ≥98.0% (by GC)
3',5'-Dichloro-2'-hydroxyacetophenone
Formula: C₈H₆Cl₂O₂
Boiling Pt: 134°C
MDL Number: MFCD00016417
CAS Number: 3321-92-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR