4-(Heptyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid

H1566-1G H1566-5G
TCH1566-1GEA 80 USD
TCH1566-1G TCH1566-5G
4-(Heptyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Heptyloxy)phenylboronic acid
Formula: C₁₃H₂₁BO₃
MDL Number: MFCD04039169
CAS Number: 136370-19-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR