3-Isopropoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Isopropoxybenzeneboronic acid
I0984-1G
I0984-5G
TCI0984-1GEA
51
USD
TCI0984-1G
TCI0984-5G
3-Isopropoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Isopropoxyphenylboronic acid
Formula:
C₉H₁₃BO₃ |
MDL Number:
MFCD03701512 CAS Number: 216485-86-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...