3-Carboxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid

C2945-1G C2945-5G
TCC2945-1GEA 71 USD
TCC2945-1G TCC2945-5G
3-Carboxy-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Carboxy-4-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MDL Number: MFCD03095129
CAS Number: 872460-12-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR