4-Carboxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Carboxy-2-fluorobenzeneboronic acid

C3220-1G C3220-5G
TCC3220-1GEA 85 USD
TCC3220-1G TCC3220-5G
4-Carboxy-2-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-2-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MDL Number: MFCD08458473
CAS Number: 851135-07-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR