3-(Ethoxycarbonyl)-4-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1187-1G E1187-5G
TCE1187-1GEA 87 USD
TCE1187-1G TCE1187-5G
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-(Ethoxycarbonyl)-4-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MDL Number: MFCD08061836
CAS Number: 874219-36-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR