3-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (3-Fluoro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 3-Fluoro-4-methoxycarbonylphenylboronic acid, 3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid

F1087-1G F1087-5G
TCF1087-1GEA 105 USD
TCF1087-1G TCF1087-5G
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
MDL Number: MFCD04115665
CAS Number: 505083-04-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR