4-(Ethoxycarbonyl)-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

E1207-1G E1207-5G
TCE1207-1GEA 85 USD
TCE1207-1G TCE1207-5G
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-(Ethoxycarbonyl)-3-fluorophenylboronic acid
Formula: C₉H₁₀BFO₄
MDL Number: MFCD06656274
CAS Number: 874288-38-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR