2-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid
F1071-1G
F1071-5G
TCF1071-1GEA
69
USD
TCF1071-1G
TCF1071-5G
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid
Formula:
C₈H₈BFO₄ MW: 197.96 g/mol Melting Pt: 140°C |
MDL Number:
MFCD08436034 CAS Number: 603122-84-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...