5-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-5-carboxyphenylboronic acid

C3246-1G C3246-5G
TCC3246-1GEA 84 USD
TCC3246-1G TCC3246-5G
5-Carboxy-2-chlorophenylboronic acid (contains varying amounts of Anhydride)
5-Carboxy-2-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MDL Number: MFCD08689491
CAS Number: 913835-75-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR