2-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3224-1G C3224-5G
TCC3224-1GEA 59 USD
TCC3224-1G TCC3224-5G
2-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-(4-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MDL Number: MFCD08689530
CAS Number: 603122-80-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR