3-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

T3162-1G T3162-5G
TCT3162-1GEA 23 USD
TCT3162-1G TCT3162-5G
3-Carboxyphenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-Carboxyphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BO₄
MDL Number: MFCD03411930
CAS Number: 269409-73-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR