2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

E1311-1G E1311-5G
TCE1311-1GEA 51 USD
TCE1311-1G TCE1311-5G
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC)
2-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
MDL Number: MFCD03453660
CAS Number: 269409-99-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 63.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR