4-Methoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (4-Methoxy-3-(trifluoromethyl)phenyl)boronic acid, 4-Methoxy-3-trifluoromethylphenylboronic acid, 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

M2839-1G M2839-200MG
TCM2839-1GEA 289 USD
TCM2839-1G TCM2839-200MG
4-Methoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Methoxy-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD07363790
CAS Number: 149507-36-8

Order Now

Specification Test Results

Appearance White to Almost white crystal to powder
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR