3-Methoxy-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid
M2943-1G
M2943-5G
TCM2943-1GEA
79
USD
TCM2943-1G
TCM2943-5G
3-Methoxy-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Methoxy-5-(trifluoromethyl)phenylboronic acid
Formula:
C₈H₈BF₃O₃ |
MDL Number:
MFCD08056362 CAS Number: 871332-97-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...