5-Hydroxy-o-toluic acid ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Hydroxy-2-methylbenzoic acid

H1586-1G H1586-5G
TCH1586-1GEA 39 USD
TCH1586-1G TCH1586-5G
5-Hydroxy-o-toluic acid ≥98.0% (by GC)
5-Hydroxy-o-toluic acid
Formula: C₈H₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09833184
CAS Number: 578-22-3

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 184.0 to 188.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR