4-Bromo-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B4843-1G
B4843-5G
TCB4843-1GEA
56
USD
TCB4843-1G
TCB4843-5G
4-Bromo-2-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-2-(trifluoromethyl)benzoic acid
Formula:
C₈H₄BrF₃O₂ |
MDL Number:
MFCD07787468 CAS Number: 320-31-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...