3-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

B5178-25G B5178-5G
TCB5178-25GEA 115 USD
TCB5178-25G TCB5178-5G
3-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
3-Bromobenzamide
Formula: C₇H₆BrNO
MW: 200.04 g/mol
MDL Number: MFCD00017127
CAS Number: 22726-00-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR