4-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T3496-1G T3496-5G
TCT3496-1GEA 41 USD
TCT3496-1G TCT3496-5G
4-(Hydroxymethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Hydroxymethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09837617
CAS Number: 302348-51-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR