HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate

A1797-25G A1797-5G
TCA1797-25GEA 428.67 USD
TCA1797-25G TCA1797-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate)
Formula: C₁₀H₁₅F₆N₆OP
Storage Temperature: Refrigerator
MDL Number: MFCD00274639
CAS Number: 148893-10-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR