6-Chlorochromone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3039-25G C3039-5G
TCC3039-25GEA 400.7 USD
TCC3039-25G TCC3039-5G
6-Chlorochromone ≥98.0% (by GC)
6-Chlorochromone
Formula: C₉H₅ClO₂
MDL Number: MFCD00191904
CAS Number: 33533-99-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR