3-Fluoro-4-nitrobenzaldehyde ≥97.0% (by GC)

Supplier: TCI America
Warning

F1130-1G F1130-5G
TCF1130-1GEA 84 USD
TCF1130-1G TCF1130-5G
3-Fluoro-4-nitrobenzaldehyde ≥97.0% (by GC)
3-Fluoro-4-nitrobenzaldehyde
Formula: C₇H₄NO₃F
MDL Number: MFCD00968940
CAS Number: 160538-51-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 53.0 to 57.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR