3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D4776-25G D4776-5G
TCD4776-25GEA 170 USD
TCD4776-25G TCD4776-5G
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride ≥98.0% (by GC, titration analysis)
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Formula: C₁₁H₆Cl₃NO₂
MDL Number: MFCD00052556
CAS Number: 4462-55-9
EINECS: 224-723-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR