6,8-Dichloro-3-formylchromone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6,8-Dichloro-3-formylchromone, 6,8-Dichlorochromone-3-carboxaldehyde, 6,8-Dichloro-4-oxo-4H-chromene-3-carbaldehyde, 6,8-Dichloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

D4388-1G D4388-5G
TCD4388-1GEA 93 USD
TCD4388-1G TCD4388-5G
6,8-Dichloro-3-formylchromone ≥98.0% (by GC)
6,8-Dichloro-3-formylchromone
Formula: C₁₀H₄Cl₂O₃
MW: 243.0430 g/mol
MDL Number: MFCD00051510
CAS Number: 64481-10-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR