1,3-Propylenediphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Propylenediphosphonic acid

P2304-1G P2304-5G
TCP2304-1GEA 156 USD
TCP2304-1G TCP2304-5G
1,3-Propylenediphosphonic acid ≥98.0% (by titrimetric analysis)
1,3-Propylenediphosphonic acid
Formula: C₃H₁₀O₆P₂
MW: 204.06 g/mol
MDL Number: MFCD00050066
CAS Number: 4671-82-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 170.0 to 177.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR