7-Bromobenzo[b]thiophene ≥98.0% (by GC)

Supplier: TCI America
Warning

B4829-1G B4829-200MG
TCB4829-1GEA 134 USD
TCB4829-1G TCB4829-200MG
7-Bromobenzo[b]thiophene ≥98.0% (by GC)
7-Bromobenzo[b]thiophene
Formula: C₈H₅BrS
Boiling Pt: 109°C
MDL Number: MFCD06657729
CAS Number: 1423-61-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR