5-Amino-2-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Methyl-3-(trifluoromethyl)aniline

M2659-1G M2659-5G
TCM2659-1GEA 27 USD
TCM2659-1G TCM2659-5G
5-Amino-2-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)
5-Amino-2-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175.16 g/mol
Flash Pt: 102°C
MDL Number: MFCD01631582
CAS Number: 65934-74-9

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR