2,6-Dichloronicotinonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

D4650-1G D4650-200MG
TCD4650-1GEA 63 USD
TCD4650-1G TCD4650-200MG
2,6-Dichloronicotinonitrile ≥98.0% (by GC)
2,6-Dichloronicotinonitrile
Formula: C₆H₂Cl₂N₂
MDL Number: MFCD03411341
CAS Number: 40381-90-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR