4-Chloro-8-fluoroquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

C3062-1G C3062-5G
TCC3062-1GEA 138 USD
TCC3062-1G TCC3062-5G
4-Chloro-8-fluoroquinoline ≥98.0% (by GC)
4-Chloro-8-fluoroquinoline
Formula: C₉H₅ClFN
MDL Number: MFCD00278785
CAS Number: 63010-72-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR