4-Chloro-6-fluoroquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

C2989-1G C2989-5G
TCC2989-1GEA 132.9 USD
TCC2989-1G TCC2989-5G
4-Chloro-6-fluoroquinoline ≥98.0% (by GC)
4-Chloro-6-fluoroquinoline
Formula: C₉H₅ClFN
MW: 181.60 g/mol
MDL Number: MFCD00278783
CAS Number: 391-77-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR