4-Dibenzofuranboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid

D4567-1G D4567-5G
TCD4567-1GEA 24 USD
TCD4567-1G TCD4567-5G
4-Dibenzofuranboronic acid (contains varying amounts of Anhydride)
4-Dibenzofuranboronic acid
Formula: C₁₂H₉BO₃
Melting Pt: 258°C
MDL Number: MFCD00092336
CAS Number: 100124-06-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) gt. 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR