5,6-Dibromo-1,3-benzodioxole ≥98.0% (by GC)
Supplier: TCI America
D4737-1G
D4737-5G
TCD4737-1GEA
39
USD
TCD4737-1G
TCD4737-5G
5,6-Dibromo-1,3-benzodioxole ≥98.0% (by GC)
5,6-Dibromo-1,3-benzodioxole
Formula:
C₇H₄Br₂O₂ |
MDL Number:
MFCD00094476 CAS Number: 5279-32-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...