3,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4807-25G D4807-5G
TCD4807-25GEA 215.5 USD
TCD4807-25G TCD4807-5G
3,5-Dibromo-α,α,α-trifluorotoluene ≥98.0% (by GC)
3,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
MDL Number: MFCD00526219
CAS Number: 401-84-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR